Assay Details
| Gene Symbol: tnika | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199703.1 | NP_955997.1 | 14 - 15 | 1803 | 112 | |
| XM_005163390.4 | XP_005163447.1 | 15 - 16 | 112 | |||
| XM_005163391.4 | XP_005163448.1 | 15 - 16 | 112 | |||
| XM_005163392.4 | XP_005163449.1 | 14 - 15 | 112 | |||
| GenBank mRNA | BC055134.1 | - | 14 - 15 | 1803 | 112 | |
More Information
Set Membership: |
|

tnika
Genomic Map