Assay Details
| Gene Symbol: tnika | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199703.1 | NP_955997.1 | 21 - 22 | 2858 | 91 | |
| XM_005163387.4 | XP_005163444.1 | 23 - 24 | 3237 | 91 | ||
| XM_005163388.4 | XP_005163445.1 | 23 - 24 | 3219 | 91 | ||
| XM_005163389.4 | XP_005163446.1 | 22 - 23 | 3150 | 91 | ||
| XM_005163390.4 | XP_005163447.1 | 22 - 23 | 2976 | 91 | ||
| XM_005163391.4 | XP_005163448.1 | 22 - 23 | 2958 | 91 | ||
| XM_005163392.4 | XP_005163449.1 | 21 - 22 | 2871 | 91 | ||
| XM_005163393.4 | XP_005163450.1 | 8 - 9 | 1362 | 91 | ||
| GenBank mRNA | BC055134.1 | - | 21 - 22 | 2858 | 91 | |
More Information
Set Membership: |
|

tnika
Genomic Map