Assay Details
| Gene Symbol: tnika | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199703.1 | NP_955997.1 | 24 - 25 | 3274 | 61 | |
| XM_005163387.4 | XP_005163444.1 | 26 - 27 | 3653 | 61 | ||
| XM_005163388.4 | XP_005163445.1 | 26 - 27 | 3635 | 61 | ||
| XM_005163389.4 | XP_005163446.1 | 25 - 26 | 3566 | 61 | ||
| XM_005163390.4 | XP_005163447.1 | 25 - 26 | 3392 | 61 | ||
| XM_005163391.4 | XP_005163448.1 | 25 - 26 | 3374 | 61 | ||
| XM_005163392.4 | XP_005163449.1 | 24 - 25 | 3287 | 61 | ||
| XM_005163393.4 | XP_005163450.1 | 11 - 12 | 1778 | 61 | ||
| GenBank mRNA | BC055134.1 | - | 24 - 25 | 3274 | 61 | |
More Information
Set Membership: |
|

tnika
Genomic Map