Assay Details
| Gene Symbol: cnn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199753.1 | NP_956047.1 | 1 - 2 | 193 | 82 | |
| GenBank mRNA | BC053309.1 | - | 1 - 2 | 193 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cnn3a| Gene Symbol: cnn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199753.1 | NP_956047.1 | 1 - 2 | 193 | 82 | |
| GenBank mRNA | BC053309.1 | - | 1 - 2 | 193 | 82 | |
Set Membership: |
|