Assay Details
| Gene Symbol: ptpn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199846.1 | NP_956140.1 | 14 - 15 | 1909 | 113 | |
| GenBank mRNA | BC045328.1 | - | 14 - 15 | 1909 | 113 | |
| BC164416.1 | - | 13 - 14 | 1749 | 113 | ||
More Information
Set Membership: |
|

ptpn11a
Genomic Map