Assay Details
| Gene Symbol: itm2cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199980.1 | NP_956274.1 | 3 - 4 | 499 | 69 | |
| XM_021479096.1 | XP_021334771.1 | 4 - 5 | 692 | 69 | ||
| GenBank mRNA | BC054689.1 | - | 3 - 4 | 499 | 69 | |
| BC164593.1 | - | 3 - 4 | 456 | 69 | ||
More Information
Set Membership: |
|

itm2cb
Genomic Map