Assay Details
| Gene Symbol: sept5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199988.1 | NP_956282.1 | 7 - 8 | 666 | 93 | |
| GenBank mRNA | BC059564.1 | - | 7 - 8 | 666 | 93 | |
More Information
Set Membership: |
|

sept5a
Genomic Map