Assay Details
| Gene Symbol: tia1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200182.2 | NP_956476.1 | 10 - 11 | 999 | 92 | |
| GenBank mRNA | AY398424.1 | - | 10 - 11 | 1066 | 92 | |
| BC045485.1 | - | 10 - 11 | 1038 | 92 | ||
| BC066734.1 | - | 10 - 11 | 999 | 92 | ||
| BC155269.1 | - | 10 - 11 | 1002 | 92 | ||
More Information
Set Membership: |
|

tia1l
Genomic Map