Assay Details
| Gene Symbol: tia1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200182.2 | NP_956476.1 | 9 - 10 | 864 | 108 | |
| XM_005165110.4 | XP_005165167.1 | 9 - 10 | 921 | 108 | ||
| GenBank mRNA | AY398424.1 | - | 9 - 10 | 931 | 108 | |
| BC045485.1 | - | 9 - 10 | 903 | 108 | ||
| BC066734.1 | - | 9 - 10 | 864 | 108 | ||
| BC155269.1 | - | 9 - 10 | 867 | 108 | ||
More Information
Set Membership: |
|

tia1l
Genomic Map