Assay Details
| Gene Symbol: smu1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200199.1 | NP_956493.1 | 10 - 11 | 1431 | 84 | |
| GenBank mRNA | BC045945.1 | - | 10 - 11 | 1431 | 84 | |
More Information
Set Membership: |
|

smu1a
Genomic Map