Assay Details
| Gene Symbol: smu1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200199.1 | NP_956493.1 | 7 - 8 | 1008 | 106 | |
| GenBank mRNA | BC045945.1 | - | 7 - 8 | 1008 | 106 | |
More Information
Set Membership: |
|

smu1a
Genomic Map