Assay Details
| Gene Symbol: ccdc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200241.2 | NP_956535.2 | 8 - 9 | 1176 | 64 | |
| GenBank mRNA | BC047810.1 | - | 8 - 9 | 1308 | 64 | |
| BC124685.1 | - | 8 - 9 | 1172 | 64 | ||
More Information
Set Membership: |
|

ccdc6b
Genomic Map