Assay Details
| Gene Symbol: ggt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200415.1 | NP_956709.1 | 13 - 14 | 1797 | 78 | |
| XM_017357293.2 | XP_017212782.1 | 13 - 14 | 2955 | 78 | ||
| GenBank mRNA | BC054631.1 | - | 13 - 14 | 1797 | 78 | |
More Information
Set Membership: |
|

ggt1a
Genomic Map