Assay Details
| Gene Symbol: ilk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200571.1 | NP_956865.1 | 10 - 11 | 1214 | 62 | |
| GenBank mRNA | BC056593.1 | - | 10 - 11 | 1214 | 62 | |
More Information
Set Membership: |
|

ilk
Genomic Map