Assay Details
| Gene Symbol: mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200736.1 | NP_957030.1 | 1 - 2 | 140 | 117 | |
| XM_005161991.4 | XP_005162048.1 | 1 - 2 | 245 | 117 | ||
| GenBank mRNA | AY398315.1 | - | 1 - 2 | 154 | 117 | |
| BC059520.1 | - | 1 - 2 | 140 | 117 | ||
More Information
Set Membership: |
|

mrgbp
Genomic Map