Assay Details
| Gene Symbol: mrgbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200736.1 | NP_957030.1 | 2 - 3 | 266 | 119 | |
| XM_005161991.4 | XP_005162048.1 | 2 - 3 | 371 | 119 | ||
| GenBank mRNA | AY398315.1 | - | 2 - 3 | 280 | 119 | |
| BC059520.1 | - | 2 - 3 | 266 | 119 | ||
More Information
Set Membership: |
|

mrgbp
Genomic Map