Assay Details
| Gene Symbol: dusp8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200880.1 | NP_957174.1 | 5 - 6 | 888 | 65 | |
| XM_005170679.4 | XP_005170736.1 | 5 - 6 | 65 | |||
| XM_005170680.4 | XP_005170737.1 | 3 - 4 | 65 | |||
| GenBank mRNA | BC063941.1 | - | 5 - 6 | 888 | 65 | |
More Information
Set Membership: |
|

dusp8a
Genomic Map