Assay Details
| Gene Symbol: b4galt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200938.1 | NP_957232.1 | 5 - 6 | 866 | 61 | |
| XM_005170141.4 | XP_005170198.1 | 5 - 6 | 1236 | 61 | ||
| GenBank mRNA | BC046890.1 | - | 5 - 6 | 866 | 61 | |
| BC164700.1 | - | 5 - 6 | 615 | 61 | ||
More Information
Set Membership: |
|

b4galt6
Genomic Map