Assay Details
| Gene Symbol: gpc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201020.1 | NP_957314.1 | 7 - 8 | 1405 | 70 | |
| GenBank mRNA | BC053161.1 | - | 7 - 8 | 1405 | 70 | |
| KC836777.1 | - | 7 - 8 | 1238 | 70 | ||
| KM655718.1 | - | 7 - 8 | 1292 | 70 | ||
More Information
Set Membership: |
|

gpc1b
Genomic Map