Assay Details
| Gene Symbol: bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201106.1 | NP_957400.1 | 4 - 5 | 715 | 57 | |
| GenBank mRNA | BC053134.1 | - | 4 - 5 | 715 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bysl| Gene Symbol: bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201106.1 | NP_957400.1 | 4 - 5 | 715 | 57 | |
| GenBank mRNA | BC053134.1 | - | 4 - 5 | 715 | 57 | |
Set Membership: |
|