Assay Details
| Gene Symbol: scamp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201194.2 | NP_957488.1 | 4 - 5 | 403 | 81 | |
| GenBank mRNA | BC055274.1 | - | 4 - 5 | 403 | 81 | |
| BC056762.1 | - | 4 - 5 | 327 | 81 | ||
More Information
Set Membership: |
|

scamp2l
Genomic Map