Assay Details
| Gene Symbol: yars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201316.1 | NP_958473.1 | 7 - 8 | 745 | 66 | |
| XM_005159598.4 | XP_005159655.1 | 7 - 8 | 770 | 66 | ||
| XM_021468004.1 | XP_021323679.1 | 7 - 8 | 736 | 66 | ||
| GenBank mRNA | AY398430.1 | - | 7 - 8 | 745 | 66 | |
More Information
Set Membership: |
|

yars1
Genomic Map