Assay Details
| Gene Symbol: hccsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201451.1 | NP_958859.1 | 5 - 6 | 608 | 78 | |
| GenBank mRNA | AY394970.1 | - | 5 - 6 | 608 | 78 | |
| BC044486.1 | - | 5 - 6 | 617 | 78 | ||
| BC065872.1 | - | 5 - 6 | 603 | 78 | ||
| BC164615.1 | - | 4 - 5 | 560 | 78 | ||
More Information
Set Membership: |
|

hccsb
Genomic Map