Assay Details
| Gene Symbol: calr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201465.3 | NP_958873.3 | 1 - 2 | 195 | 97 | |
| GenBank mRNA | BC046906.1 | - | 1 - 2 | 158 | 97 | |
| BC075778.1 | - | 1 - 2 | 180 | 97 | ||
| KM655575.1 | - | 1 - 2 | 98 | 97 | ||
More Information
Set Membership: |
|

calr3b
Genomic Map