Assay Details
| Gene Symbol: pfn2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201466.2 | NP_958874.2 | 2 - 3 | 489 | 88 | |
| GenBank mRNA | BC078650.1 | - | 2 - 3 | 489 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pfn2l| Gene Symbol: pfn2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201466.2 | NP_958874.2 | 2 - 3 | 489 | 88 | |
| GenBank mRNA | BC078650.1 | - | 2 - 3 | 489 | 88 | |
Set Membership: |
|