Assay Details
| Gene Symbol: arf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201480.1 | NP_958888.1 | 2 - 3 | 236 | 95 | |
| GenBank mRNA | AY577008.1 | - | 2 - 3 | 251 | 95 | |
| BC046063.1 | - | 2 - 3 | 236 | 95 | ||
| BC062853.1 | - | 2 - 3 | 204 | 95 | ||
More Information
Set Membership: |
|

arf2b
Genomic Map