Assay Details
| Gene Symbol: mcm3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171068.1 | NP_001164539.1 | 2 - 3 | 305 | 122 | |
| GenBank mRNA | BC045431.1 | - | 2 - 3 | 293 | 122 | |
More Information
Set Membership: |
|

mcm3l
Genomic Map