Assay Details
| Gene Symbol: mcm3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB973819.1 | - | 8 - 9 | 1160 | 69 | |
| BC045431.1 | - | 8 - 9 | 1267 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mcm3l| Gene Symbol: mcm3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB973819.1 | - | 8 - 9 | 1160 | 69 | |
| BC045431.1 | - | 8 - 9 | 1267 | 69 | ||
Set Membership: |
|