Assay Details
| Gene Symbol: mcm3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171068.1 | NP_001164539.1 | 9 - 10 | 1483 | 86 | |
| GenBank mRNA | AB973819.1 | - | 9 - 10 | 1364 | 86 | |
| BC045431.1 | - | 9 - 10 | 1471 | 86 | ||
More Information
Set Membership: |
|

mcm3l
Genomic Map