Assay Details
| Gene Symbol: scamp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205547.1 | NP_991110.1 | 4 - 5 | 388 | 84 | |
| GenBank mRNA | BC065670.1 | - | 4 - 5 | 388 | 84 | |
More Information
Set Membership: |
|

scamp5a
Genomic Map