Assay Details
| Gene Symbol: lman2la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205725.2 | NP_991288.2 | 4 - 5 | 657 | 72 | |
| GenBank mRNA | AF506214.1 | - | 4 - 5 | 692 | 72 | |
| BC097031.1 | - | 4 - 5 | 657 | 72 | ||
More Information
Set Membership: |
|

lman2la
Genomic Map