Assay Details
| Gene Symbol: dgcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205739.1 | NP_991302.1 | 4 - 5 | 508 | 84 | |
| XM_017357308.2 | XP_017212797.1 | 4 - 5 | 601 | 84 | ||
| GenBank mRNA | AY398335.1 | - | 4 - 5 | 508 | 84 | |
More Information
Set Membership: |
|

dgcr6
Genomic Map