Assay Details
| Gene Symbol: kcnh6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212837.1 | NP_998002.1 | 4 - 5 | 666 | 78 | |
| XM_005164007.4 | XP_005164064.1 | 4 - 5 | 1136 | 78 | ||
| XM_005164008.4 | XP_005164065.1 | 4 - 5 | 1142 | 78 | ||
| GenBank mRNA | AF532865.1 | - | 4 - 5 | 666 | 78 | |
| BC163642.1 | - | 4 - 5 | 633 | 78 | ||
More Information
Set Membership: |
|

kcnh6a
Genomic Map