Assay Details
| Gene Symbol: runx2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212862.2 | NP_998027.2 | 4 - 5 | 650 | 89 | |
| XM_021468385.1 | XP_021324060.1 | 4 - 5 | 1317 | 89 | ||
| GenBank mRNA | AY176052.1 | - | 3 - 4 | 621 | 89 | |
| AY382180.1 | - | 4 - 5 | 616 | 89 | ||
| AY382181.1 | - | 3 - 4 | 630 | 89 | ||
| AY443096.1 | - | 4 - 5 | 652 | 89 | ||
| AY443097.1 | - | 3 - 4 | 649 | 89 | ||
| AY817150.1 | - | 2 - 3 | 244 | 89 | ||
More Information
Set Membership: |
|

runx2b
Genomic Map