Assay Details
| Gene Symbol: lgals2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212894.2 | NP_998059.2 | 2 - 3 | 161 | 113 | |
| GenBank mRNA | AY421704.1 | - | 2 - 3 | 131 | 113 | |
| BC071424.1 | - | 2 - 3 | 127 | 113 | ||
| EH454585.1 | - | 2 - 3 | 157 | 113 | ||
More Information
Set Membership: |
|

lgals2a
Genomic Map