Assay Details
| Gene Symbol: wdr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212946.1 | NP_998111.2 | 3 - 4 | 887 | 70 | |
| GenBank mRNA | BC067591.1 | - | 3 - 4 | 887 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdr32| Gene Symbol: wdr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212946.1 | NP_998111.2 | 3 - 4 | 887 | 70 | |
| GenBank mRNA | BC067591.1 | - | 3 - 4 | 887 | 70 | |
Set Membership: |
|