Assay Details
| Gene Symbol: wdr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212946.1 | NP_998111.2 | 5 - 6 | 1176 | 87 | |
| GenBank mRNA | BC067591.1 | - | 5 - 6 | 1176 | 87 | |
More Information
Set Membership: |
|

wdr32
Genomic Map