Assay Details
| Gene Symbol: pak5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212962.1 | NP_998127.1 | 3 - 4 | 1723 | 58 | |
| GenBank mRNA | BC068016.1 | - | 3 - 4 | 1723 | 58 | |
| BC168506.1 | - | 3 - 4 | 1618 | 58 | ||
More Information
Set Membership: |
|

pak5
Genomic Map