Assay Details
| Gene Symbol: ggctb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213005.1 | NP_998170.1 | 1 - 2 | 79 | 95 | |
| GenBank mRNA | BC045940.1 | - | 1 - 2 | 79 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ggctb| Gene Symbol: ggctb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213005.1 | NP_998170.1 | 1 - 2 | 79 | 95 | |
| GenBank mRNA | BC045940.1 | - | 1 - 2 | 79 | 95 | |
Set Membership: |
|