Assay Details
| Gene Symbol: ggctb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213005.1 | NP_998170.1 | 2 - 3 | 229 | 90 | |
| XM_005173544.3 | XP_005173601.1 | 2 - 3 | 247 | 90 | ||
| XM_021466669.1 | XP_021322344.1 | 2 - 3 | 268 | 90 | ||
| XM_021466670.1 | XP_021322345.1 | 2 - 3 | 348 | 90 | ||
| GenBank mRNA | BC045940.1 | - | 2 - 3 | 229 | 90 | |
More Information
Set Membership: |
|

ggctb
Genomic Map