Assay Details
| Gene Symbol: hspa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213058.1 | NP_998223.1 | 8 - 9 | 1534 | 67 | |
| GenBank mRNA | AY648749.1 | - | 8 - 9 | 1537 | 67 | |
| BC052971.1 | - | 8 - 9 | 1534 | 67 | ||
| BC063946.1 | - | 8 - 9 | 1515 | 67 | ||
More Information
Set Membership: |
|

hspa5
Genomic Map