Assay Details
| Gene Symbol: tcirg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213069.1 | NP_998234.1 | 7 - 8 | 829 | 76 | |
| GenBank mRNA | BC045484.1 | - | 7 - 8 | 829 | 76 | |
| BC165671.1 | - | 6 - 7 | 742 | 76 | ||
More Information
Set Membership: |
|

tcirg1b
Genomic Map