Assay Details
| Gene Symbol: ptges3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213170.1 | NP_998335.1 | 2 - 3 | 285 | 118 | |
| GenBank mRNA | AY724692.1 | - | 1 - 2 | 115 | 118 | |
| BC049454.1 | - | 2 - 3 | 208 | 118 | ||
| BC056294.1 | - | 2 - 3 | 276 | 118 | ||
| BC065314.1 | - | 2 - 3 | 285 | 118 | ||
| BC165135.1 | - | 1 - 2 | 149 | 118 | ||
More Information
Set Membership: |
|

ptges3a
Genomic Map