Assay Details
| Gene Symbol: ptges3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213170.1 | NP_998335.1 | 3 - 4 | 356 | 150 | |
| GenBank mRNA | AY724692.1 | - | 2 - 3 | 186 | 150 | |
| BC049454.1 | - | 3 - 4 | 279 | 150 | ||
| BC056294.1 | - | 3 - 4 | 347 | 150 | ||
| BC065314.1 | - | 3 - 4 | 356 | 150 | ||
| BC165135.1 | - | 2 - 3 | 220 | 150 | ||
More Information
Set Membership: |
|

ptges3a
Genomic Map