Assay Details
| Gene Symbol: ptges3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213170.1 | NP_998335.1 | 5 - 6 | 542 | 78 | |
| GenBank mRNA | AY724692.1 | - | 4 - 5 | 372 | 78 | |
| BC056294.1 | - | 5 - 6 | 533 | 78 | ||
| BC065314.1 | - | 5 - 6 | 542 | 78 | ||
More Information
Set Membership: |
|

ptges3a
Genomic Map