Assay Details
| Gene Symbol: p4hb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213364.3 | NP_998529.3 | 2 - 3 | 505 | 87 | |
| GenBank mRNA | EB796102.1 | - | 2 - 3 | 250 | 87 | |
| EB848925.1 | - | 2 - 3 | 509 | 87 | ||
More Information
Set Membership: |
|

p4hb
Genomic Map