Assay Details
| Gene Symbol: pdha1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213393.1 | NP_998558.1 | 9 - 10 | 1027 | 105 | |
| XM_005165134.4 | XP_005165191.1 | 10 - 11 | 1013 | 105 | ||
| GenBank mRNA | BC060928.1 | - | 9 - 10 | 940 | 105 | |
| BC071373.1 | - | 9 - 10 | 1027 | 105 | ||
More Information
Set Membership: |
|

pdha1a
Genomic Map