Assay Details
| Gene Symbol: kctd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213478.2 | NP_998643.1 | 6 - 7 | 795 | 93 | |
| GenBank mRNA | BC061457.1 | - | 6 - 7 | 767 | 93 | |
More Information
Set Membership: |
|

kctd10
Genomic Map