Assay Details
| Gene Symbol: mfsd14a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213527.2 | NP_998692.2 | 7 - 8 | 1005 | 72 | |
| GenBank mRNA | BC045310.1 | - | 7 - 8 | 1003 | 72 | |
| BC097075.1 | - | 7 - 8 | 999 | 72 | ||
More Information
Set Membership: |
|

mfsd14a1
Genomic Map