Assay Details
| Gene Symbol: pgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213453.2 | NP_998618.2 | 9 - 10 | 1214 | 65 | |
| GenBank mRNA | AY391449.1 | - | 9 - 10 | 1186 | 65 | |
| BC044196.1 | - | 9 - 10 | 1214 | 65 | ||
| BC095571.1 | - | 9 - 10 | 1179 | 65 | ||
| BC100123.1 | - | 9 - 10 | 1180 | 65 | ||
| BC165755.1 | - | 9 - 10 | 1229 | 65 | ||
More Information
Set Membership: |
|

pgd
Genomic Map